Abnova
Manufacturer Code:H00005264B01
Catalog # 89-005-956
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | PHYH (NP_006205 1 a.a. ~ 338 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human PHYH protein.
For Research Use Only
Protein Aliases: LN1; LNAP1; PAHX; PhyH; PHYH1; Phytanic acid oxidase; phytanoil-CoA alpha hydroxylase; phytanoyl-CoA 2 oxoglutarate dioxygenase; Phytanoyl-CoA alpha-hydroxylase; Phytanoyl-CoA dioxygenase, peroxisomal; RD
Gene Aliases: LN1; LNAP1; PAHX; PHYH; PHYH1; RD
UniProt ID: (Human) O14832
Entrez Gene ID: (Human) 5264
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.