Abnova
Manufacturer Code:PAB8516
Catalog # 89-133-717
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 12-22 at N-terminus of arabidopsis phototropin 2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of phototropin 2.
For Research Use Only
Protein Aliases: AtKin7; Defective in chloroplast avoidance protein 1; K21L19.6; K21L19_6; NON PHOTOTROPIC HYPOCOTYL 1-LIKE; Non-phototropic hypocotyl 1-like protein 1; NPL1; Phototropin-2
Gene Aliases: AT5G58140; AtPHOT2; CAV1; K21L19.6; K21L19_6; KIN7; NON PHOTOTROPIC HYPOCOTYL 1-LIKE; NPL1; PHOT2; phototropin 2
UniProt ID: (Arabidopsis) P93025
Entrez Gene ID: (Arabidopsis) 835926
Molecular Function:
annexin
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
kinase
non-receptor serine/threonine protein kinase
protein kinase
transfer/carrier protein
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.