Abnova
Manufacturer Code:PAB8518
Catalog # 89-133-718
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to arabidopsis phototropin 1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of phototropin 1.
For Research Use Only
Protein Aliases: F16L2.3; JK224; Non-phototropic hypocotyl protein 1; NONPHOTOTROPIC HYPOCOTYL 1; NPH1; PHOTOTROPIN; phototropin 1; Phototropin-1; putative serine/threonine protein kinase; ROOT PHOTOTROPISM 1; Root phototropism protein 1; RPT1
Gene Aliases: AT3G45780; F16L2.3; JK224; NONPHOTOTROPIC HYPOCOTYL 1; NPH1; PHOT1; PHOTOTROPIN; phototropin 1; ROOT PHOTOTROPISM 1; RPT1; T6D9_110
UniProt ID: (Arabidopsis) O48963
Entrez Gene ID: (Arabidopsis) 823721
Molecular Function:
annexin
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
kinase
non-receptor serine/threonine protein kinase
protein kinase
transfer/carrier protein
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.