Abnova
Manufacturer Code:H00023236A01
Catalog # 89-018-862
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | PLCB1 (NP_056007 1107 a.a. ~ 1216 a.a) partial recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant PLCB1.
For Research Use Only
Protein Aliases: 1-phosphatidyl-D-myo-inositol-4,5-bisphosphate; 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase beta-1; FLJ45792; inositoltrisphosphohydrolase; monophosphatidylinositol phosphodiesterase; phosphoinositidase C; Phosphoinositide phospholipase C-beta-1; phospholipase C, beta 1 (phosphoinositide-specific); Phospholipase C-beta-1; Phospholipase C-I; PI-PLC; PLC-154; PLC-beta-1; PLC-I; PLC154; triphosphoinositide phosphodiesterase
Gene Aliases: EIEE12; KIAA0581; PI-PLC; PLC-154; PLC-beta-1; PLC-I; PLC154; PLCB1; PLCB1A; PLCB1B
UniProt ID: (Human) Q9NQ66
Entrez Gene ID: (Human) 23236
Molecular Function:
G-protein modulator
calcium-binding protein
enzyme modulator
guanyl-nucleotide exchange factor
hydrolase
lipase
phospholipase
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.