Abnova
Manufacturer Code:H00009468A01
Catalog # 89-004-441
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | PCYT1B (NP_004836 236 a.a. ~ 314 a.a) partial recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant PCYT1B.
For Research Use Only
Protein Aliases: CCT B; CCT-beta; Choline-phosphate cytidylyltransferase B; CT B; CTB; CTP:phosphocholine cytidylyltransferase B; Phosphorylcholine transferase B
Gene Aliases: CCTB; CTB; PCYT1B
UniProt ID: (Human) Q9Y5K3
Entrez Gene ID: (Human) 9468
Molecular Function: nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.