Abnova
Manufacturer Code:H00005191A01
Catalog # 89-004-234
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | PEX7 (NP_000279 2 a.a. ~ 99 a.a) partial recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant PEX7.
For Research Use Only
Protein Aliases: Peroxin-7; peroxisomal PTS2 receptor; Peroxisomal targeting signal 2 receptor; peroxisome targeting signal 2 receptor; PTS2 receptor; PTS2R; RCDP1; RD
Gene Aliases: PBD9B; PEX7; PTS2R; RCDP1; RD
UniProt ID: (Human) O00628
Entrez Gene ID: (Human) 5191
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.