Abnova
Manufacturer Code:H00005824B01P
Catalog # 89-016-893
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | PEX19 (NP_002848.1 1 a.a. ~ 299 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human PEX19 protein.
For Research Use Only
Protein Aliases: 33 kDa housekeeping protein; D1S2223E; HK33; housekeeping gene, 33kD; Peroxin-19; Peroxisomal biogenesis factor 19; Peroxisomal farnesylated protein; PMP1; PMPI; PXF; PXMP1
Gene Aliases: D1S2223E; HK33; OK/SW-cl.22; PBD12A; PEX19; PMP1; PMPI; PXF; PXMP1
UniProt ID: (Human) P40855
Entrez Gene ID: (Human) 5824
Molecular Function:
storage protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.