Abnova
Manufacturer Code:H00010455B01P
Catalog # 89-017-002
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | PECI (NP_006108.2 1 a.a. ~ 364 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human PECI protein.
For Research Use Only
Protein Aliases: ACBD2; acyl-Coenzyme A binding domain containing 2; D3,D2-enoyl-CoA isomerase; DBI-related protein 1; Delta(3),delta(2)-enoyl-CoA isomerase; Diazepam-binding inhibitor-related protein 1; dJ1013A10.3; Dodecenoyl-CoA isomerase; DRS-1; DRS1; Enoyl-CoA delta isomerase 2; HCA88; Hepatocellular carcinoma-associated antigen 88; KIAA0536; pECI; Peroxisomal 3,2-trans-enoyl-CoA isomerase; peroxisomal D3,D2-enoyl-CoA isomerase; Renal carcinoma antigen NY-REN-1; testicular secretory protein Li 33
Gene Aliases: ACBD2; dJ1013A10.3; DRS-1; DRS1; ECI2; HCA88; PECI
UniProt ID: (Human) O75521
Entrez Gene ID: (Human) 10455
Molecular Function:
enzyme modulator
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.