Abnova
Manufacturer Code:H00005478M01
Catalog # 89-014-173
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1F4-1B5 |
Immunogen | PPIA (AAH00689.1 1 a.a. ~ 165 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant PPIA.
For Research Use Only
Protein Aliases: Cyclophilin A; Cyclosporin A-binding protein; CYPA; CYPH; epididymis secretory sperm binding protein Li 69p; MGC117158; MGC12404; MGC23397; Peptidyl-prolyl cis-trans isomerase A; Peptidyl-prolyl cis-trans isomerase A, N-terminally processed; peptidylprolyl isomerase A (cyclophilin A); PPIase A; Rotamase A; T cell cyclophilin
Gene Aliases: CYPA; CYPH; HEL-S-69p; PPIA
UniProt ID: (Human) P62937
Entrez Gene ID: (Human) 5478
Molecular Function: isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.