Proteintech
Manufacturer Code:242941AP150UL
Catalog # 50-173-4282
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | peptide YY Fusion Protein Ag17939 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Peptide tyrosine tyrosine; Peptide tyrosine tyrosine peptide YY PYY PYY I PYY II PYY1; Peptide YY; Peptide YY(3-36); prepro-PYY; PYY; PYY-I; PYY-II
Gene Aliases: PYY; PYY-I; PYY1
UniProt ID: (Human) P10082
Entrez Gene ID: (Human) 5697
Molecular Function: neuropeptide peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.