Abnova
Manufacturer Code:H00005806M01
Catalog # 89-000-923
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5B7 |
Immunogen | PTX3 (NP_002843 282 a.a. ~ 380 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant PTX3.
For Research Use Only
Protein Aliases: long pentraxin 3; Pentaxin-related protein PTX3; Pentraxin-related protein PTX3; TNF alpha-induced protein 5; TNFAIP5; TSG-14; Tumor necrosis factor alpha-induced protein 5; Tumor necrosis factor-inducible gene 14 protein; tumor necrosis factor-inducible protein TSG-14
Gene Aliases: PTX3; TNFAIP5; TSG-14; TSG14
UniProt ID: (Human) P26022
Entrez Gene ID: (Human) 5806
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.