Invitrogen
Manufacturer Code:PA599123
Catalog # PIPA599123
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit human protein purified from E.coli (HIS-beta-Synuclein) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-synuclein; Beta-synuclein; Breast cancer-specific gene 1 protein; Gamma-synuclein; NACP; non A-beta component of AD amyloid; Non-A beta component of AD amyloid; non-A4 component of amyloid; Non-A4 component of amyloid precursor; Persyn; Phosphoneuroprotein 14; PNP 14; Sensory neuron synuclein; SR; Synoretin; synuclein alpha-140; synuclein, alpha (non A4 component of amyloid precursor); synuclein, gamma (breast cancer-specific protein 1)
Gene Aliases: AI838531; alphaSYN; BCSG1; betaSYN; C79089; NACP; PARK1; PARK4; PD1; PERSYN; PRSN; SNCA; SNCB; SNCG; SR; Syn
UniProt ID: (Human) P37840, (Human) Q16143, (Human) O76070, (Rat) P37377, (Mouse) Q9CXF8, (Mouse) Q91ZZ3, (Rat) Q63754, (Rat) Q63544, (Mouse) Q9Z0F7
Entrez Gene ID: (Human) 6622, (Human) 6620, (Human) 6623, (Rat) 29219, (Mouse) 20617, (Mouse) 104069, (Rat) 113893, (Rat) 64347, (Mouse) 20618
Molecular Function: chaperone membrane traffic protein cytoskeletal protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.