Invitrogen
Manufacturer Code:PA532609
Catalog # PIPA532609
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide near N-terminus of human alpha beta gamma synuclein protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-synuclein; Beta-synuclein; Breast cancer-specific gene 1 protein; Gamma-synuclein; NACP; non A-beta component of AD amyloid; Non-A beta component of AD amyloid; Non-A4 component of amyloid precursor; PD1 NACP PARK1 PARK4 SR BCSG1; Persyn; SR; Synoretin; synuclein alpha-140; synuclein, alpha (non A4 component of amyloid precursor); synuclein, gamma (breast cancer-specific protein 1)
Gene Aliases: BCSG1; NACP; PARK1; PARK4; PD1; PERSYN; PRSN; SNCA; SNCB; SNCG; SR
UniProt ID: (Human) P37840, (Human) Q16143, (Human) O76070
Entrez Gene ID: (Human) 6622, (Human) 6620, (Human) 6623
Molecular Function: chaperone membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.