Invitrogen
Manufacturer Code:MA1012
Catalog # PIMA1012
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | Ras10 |
Immunogen | Purified recombit Ras protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: c-H-ras; c-Ha-ras p21 protein; c-Ha-ras transgene; c-has/bas p21 protein; c-K-ras; c-Ki-ras; c-Kirsten-ras protein; c-ras-Ki-2 activated oncogene; cellular c-Ki-ras2 proto-oncogene; GTP- and GDP-binding peptide B; GTPase HRas; GTPase HRas, N-terminally processed; GTPase KRas; GTPase KRas, N-terminally processed; GTPase NRas; H-ras 1 protein; H-Ras-1; Ha-Ras; Ha-Ras Harvey-ras Hras H-Ras-1 Transforming protein p21 c-H-ras p21ras Ras GTPase Hras Hras1 C-K-RAS RASK K-RAS2A K-RAS2B K-RAS4A K-RAS4B KI-RAS KRAS1 KRAS2 RASK2 GTPase KRas K-Ras 2 K-ras p21 protein Kirsten rat sarcoma-2 viral (v-Ki-ras2) oncogene homolog PR310 c-K-ras oncogene c-Ki-ras c-Kirsten-ras protein cellular c-Ki-ras2 N-RAS Neuroblastoma RAS NRAS NRAS1 NS6 ALPS4 Neuroblastoma RAS viral (v-ras) oncogene homolog RAS RASN GTPase Nras Transforming protein N-Ras; Ha-Ras1 proto-oncoprotein; Harvey rat sarcoma viral oncogene homolog; Harvey rat sarcoma viral oncoprotein; Harvey rat sarcoma virus oncogene 1; K-Ras 2; K-ras p21 protein; Ki-Ras; Kirsten rat sarcoma oncogene 2, expressed; Kirsten rat sarcoma viral oncogene homolog; N-ras protein part 4; neuroblastoma RAS viral (v-ras) oncogene homolog; oncogene KRAS2; p19 H-RasIDX protein; p21 protein; p21ras; PR310 c-K-ras oncogene; Ras family small GTP binding protein H-Ras; transformation gene: oncogene HAMSV; Transforming protein N-Ras; Transforming protein p21; v-Ha-ras Harvey rat sarcoma viral oncogene homolog; v-Ki-ras2 Kirsten rat sarcoma 2 viral oncogene homolog; v-ras neuroblastoma RAS viral oncogene homolog
Gene Aliases: AI929937; ALPS4; AV095280; C-BAS/HAS; C-H-RAS; c-Ha-ras; C-HA-RAS1; C-K-RAS; c-rasHa; CFC2; CMNS; CTLO; H-ras; H-RASIDX; Ha-ras; HAMSV; Harvey-ras; HRAS; Hras-1; HRAS1; K-ras; K-RAS2A; K-RAS2B; K-RAS4A; K-RAS4B; KI-RAS; KRAS; Kras-2; KRAS1; KRAS2; N-ras; NCMS; NRAS; NRAS1; NS; NS3; NS6; p21B; p21ras; RALD; ras; RASH1; RASK2
UniProt ID: (Human) P01112, (Mouse) Q61411, (Human) P01116, (Mouse) P32883, (Human) P01111
Entrez Gene ID: (Human) 3265, (Mouse) 15461, (Human) 3845, (Mouse) 16653, (Human) 4893, (Mouse) 18176
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.