Invitrogen
Manufacturer Code:PA577740
Catalog # PIPA577740
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)TRYGKELSMVKIPSK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acid sensing (proton gated) ion channel 1; acid sensing (proton gated) ion channel 2; acid sensing (proton gated) ion channel 3; acid sensing (proton gated) ion channel family member 4; acid sensing ion channel; acid sensing ion channel 1; acid sensing ion channel 2; acid sensing ion channel 3; acid sensing ion channel family member 4; acid-sensing (proton-gated) ion channel 1; acid-sensing (proton-gated) ion channel 2; acid-sensing (proton-gated) ion channel 3; acid-sensing (proton-gated) ion channel family member 4; Acid-sensing ion channel; Acid-sensing ion channel 1; acid-sensing ion channel 1a protein; Acid-sensing ion channel 2; Acid-sensing ion channel 3; Acid-sensing ion channel 4; Acid-sensing ion channel Acid-sensing (proton-gated) ion channel ACCN ACCN2 ACCN3 ACCN4; Amiloride-sensitive brain sodium channel; Amiloride-sensitive brain sodium channel 2; Amiloride-sensitive cation channel 1, neuronal; amiloride-sensitive cation channel 1, neuronal (degenerin); Amiloride-sensitive cation channel 2, neuronal; Amiloride-sensitive cation channel 3; amiloride-sensitive cation channel 3, testis; Amiloride-sensitive cation channel 4; Amiloride-sensitive cation channel 4, pituitary; amiloride-sensitive cation channel family member 4, pituitary; Amiloride-sensitive cation channel neuronal 1; ASIC1; ASIC2; ASIC3; ASIC4; BNaC2; BNC1; Brain sodium channel 1; Brain sodium channel 2; brain sodium channel 4; Cation channel, amiloride-sensitive, neuronal, 2; degenerin; degenerin 2; Dorsal root ASIC; DRASIC; hTNaC1; Mammalian degenerin homolog; MDEG; modulatory subunit of ASIC2a; neuronal amiloride-sensitive cation channel 1; Neuronal amiloride-sensitive cation channel 3; proton gated cation channel ASIC1; proton gated cation channel DRASIC; proton-gated cation channel subunit; putative acid-sensing ion channel; Spinal chord ASIC; Testis sodium channel 1
Gene Aliases: ACCN; ACCN1; ACCN2; ACCN3; ACCN4; ACIC2; AI843610; ASIC; ASIC1; ASIC1a; ASIC2; ASIC2a; ASIC3; ASIC4; AW742291; B530003N02Rik; BNAC1; BNaC1a; BNAC2; BNAC4; BNC1; BNC1k; DRASIC; hBNaC1; MDEG; MDEG1; MDEG2; SLNAC1; Spasic; TNAC1
UniProt ID: (Human) P78348, (Human) Q16515, (Human) Q9UHC3, (Human) Q96FT7, (Mouse) Q6NXK8, (Rat) P55926, (Rat) Q62962, (Mouse) Q925H0, (Mouse) Q6X1Y6, (Rat) O35240, (Rat) Q9JHS6, (Mouse) Q7TNS7
Entrez Gene ID: (Human) 41, (Human) 40, (Human) 9311, (Human) 55515, (Mouse) 11419, (Rat) 79123, (Rat) 25364, (Mouse) 11418, (Mouse) 171209, (Rat) 286920, (Rat) 63882, (Mouse) 241118
Molecular Function:
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.