Abnova
Manufacturer Code:H00005307D01P
Catalog # 89-017-955
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PITX1 (NP_002644.3 1 a.a. ~ 314 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a full-length human PITX1 protein.
For Research Use Only
Protein Aliases: BFT; hindlimb expressed homeobox protein backfoot; Hindlimb-expressed homeobox protein backfoot; Homeobox protein PITX1; paired-like homeodomain 1; Paired-like homeodomain transcription factor 1; pituitary homeo box 1; Pituitary homeobox 1; pituitary otx-related factor; POTX; PTX1
Gene Aliases: BFT; CCF; LBNBG; PITX1; POTX; PTX1
UniProt ID: (Human) P78337
Entrez Gene ID: (Human) 5307
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.