Abnova
Manufacturer Code:H00000401B01
Catalog # 89-004-935
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | PHOX2A (NP_005160.2 1 a.a. ~ 284 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human PHOX2A protein.
For Research Use Only
Protein Aliases: aristaless homeobox homolog; Aristaless homeobox protein homolog; ARIX; arix homeodomain protein; ARIX1 homeodomain protein; CFEOM2; FEOM2; MGC52227; NCAM2; Paired mesoderm homeobox protein 2A; Paired-like homeobox 2A; PMX2A
Gene Aliases: ARIX; CFEOM2; FEOM2; NCAM2; PHOX2A; PMX2A
UniProt ID: (Human) O14813
Entrez Gene ID: (Human) 401
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.