R&D Systems
Manufacturer Code:MAB2717
Catalog # MAB2717
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 825724 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human pIgR Lys19-Arg638 Accession # CAA51532 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ22667 hepatocellular carcinoma associated protein TB6 Hepatocellular carcinoma-associated protein TB6 MGC125361 MGC125362 PIgR Poly-Ig receptor polymeric immunoglobulin receptor; hepatocellular carcinoma associated protein TB6; Hepatocellular carcinoma-associated protein TB6; PIgR; poly-Ig receptor; Polymeric immunoglobulin receptor; Secretory component
Gene Aliases: PIGR
UniProt ID: (Human) P01833
Entrez Gene ID: (Human) 5284
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.