Novus Biologicals
Manufacturer Code:NBP267008
Catalog # NBP267008
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JM11-15 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: 15S Mg(2+)-ATPase p97 subunit; 15S Mg(2+)-ATPase p97 subunit IBMPFDMGC8560 MGC131997 p97 TER ATPase TERA transitional endoplasmic reticulum ATPase valosin containing protein Valosin-containing proteinMGC148092 yeast Cdc48p homolog; epididymis luminal protein 220; epididymis secretory protein Li 70; TER ATPase; Transitional endoplasmic reticulum ATPase; Valosin-containing protein; VCP; yeast Cdc48p homolog
Gene Aliases: ALS14; CMT2Y; HEL-220; HEL-S-70; IBMPFD; IBMPFD1; p97; TERA; VCP
UniProt ID: (Human) P55072
Entrez Gene ID: (Human) 7415
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.