Invitrogen
Manufacturer Code:MA516167
Catalog # PIMA516167
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5B429 |
Immunogen | This antibody was raised against full-length human p73. It reacts with an epitope that is located on the N-terminal region of human p73. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: p53-like transcription factor; p53-related protein; P73 p53-like transcription factor p53-related protein; transformation related protein 73; Tumor protein p73
Gene Aliases: deltaNp73; P73; TAp73; TP73; Trp73
UniProt ID: (Human) O15350, (Mouse) Q9JJP2
Entrez Gene ID: (Human) 7161, (Rat) 362675, (Mouse) 22062
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.