R&D Systems
Manufacturer Code:AF1916SP
Catalog # AF1916SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human p63/TP73L Met40-Cys339 Accession # Q9H3D4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: amplified in squamous cell carcinoma; amplified in squamous cell carcinoma Chronic ulcerative stomatitis protein CUSP EEC3 EEC3B(p51B) Keratinocyte transcription factor KET KET KETRHS LMS OFC8 OFC8SHFM4TP73LNBP p40 p51 p51tumor protein p53-like p53CP P63 p63AIS p73H P73L p73LB(p51A) SHFM4 TP53CP TP53L TP63 TP73L Transformation-related protein 63 tumor protein 63 tumor protein p53-competing protein tumor protein p63 tumor protein p63 deltaN isoform delta Tumor protein p73-like; Chronic ulcerative stomatitis protein; CUSP; Keratinocyte transcription factor KET; p40; p51; p63; p73L; TP63; Transformation-related protein 63; Tumor protein 63; tumor protein p53-competing protein; Tumor protein p73-like
Gene Aliases: AIS; B(p51A); B(p51B); EEC3; KET; LMS; NBP; OFC8; p40; p51; p53CP; P63; P73H; P73L; RHS; SHFM4; TP53CP; TP53L; TP63; TP73L
UniProt ID: (Human) Q9H3D4
Entrez Gene ID: (Human) 8626
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.