Novus Biologicals
Manufacturer Code:NBP223490
Catalog # NBP223490
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5H7E2 |
Immunogen | Partial recombinant human p62/SQSTM1 (between residues 100-400) expressed in E. coli [UniProt Q13501] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: A170 EBI3-associated protein of 60 kDa EBI3-associated protein p60 EBIAP ORCA OSIL oxidative stress induced like p60PDB3 p62 p62B Paget disease of bone 3 phosphotyrosine independent ligand for the Lck SH2 domain p62 Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa sequestosome 1 sequestosome-1 Ubiquitin-binding protein p62 ZIP3; EBI3-associated protein of 60 kDa; EBI3-associated protein p60; EBIAP; oxidative stress induced like; phosphotyrosine independent ligand for the Lck SH2 domain p62; Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa; Sequestosome-1; Ubiquitin-binding protein p62
Gene Aliases: A170; FTDALS3; ORCA; OSIL; p60; p62; p62B; PDB3; SQSTM1; ZIP3
UniProt ID: (Human) Q13501
Entrez Gene ID: (Human) 8878
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.