Invitrogen
Manufacturer Code:MA534751
Catalog # PIMA534751
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JG37-64 |
Immunogen | Full length recombinant protein of human p53DINP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 2700057G22Rik p53-dependent damage-inducible nuclear protein 1 P53DINP1 SIP SIP18 SIP27 Stinp Stress-induced protein TEAP Thymus-expressed acidic protein TP53DINP1 TP53INP1 TP53INP1A TP53INP1B Trp53inp1 Tumor protein p53-inducible nuclear protein 1; p53-dependent damage-inducible nuclear protein 1; p53-inducible p53DINP1; p53DINP1; Stress-induced protein; Tumor protein p53-inducible nuclear protein 1
Gene Aliases: P53DINP1; SIP; Teap; TP53DINP1; TP53INP1; TP53INP1A; TP53INP1B
UniProt ID: (Human) Q96A56
Entrez Gene ID: (Human) 94241
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.