Invitrogen
Manufacturer Code:PA596989
Catalog # PIPA596989
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-100 of human TP53INP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: p53-dependent damage-inducible nuclear protein 1; p53-inducible p53DINP1; p53DINP1; stress induced protein; Stress-induced protein; TEAP; Thymus-expressed acidic protein; transformation related protein 53 inducible nuclear protein 1; Tumor protein p53-inducible nuclear protein 1
Gene Aliases: P53DINP1; SIP; Stinp; Teap; TP53DINP1; TP53INP1; TP53INP1A; TP53INP1B; Trp53inp1
UniProt ID: (Human) Q96A56, (Rat) Q80YE2
Entrez Gene ID: (Human) 94241, (Rat) 297822
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.