Invitrogen
Manufacturer Code:PA529430
Catalog # PIPA529430
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 1 and 59 of Human p53DINP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: p53-dependent damage-inducible nuclear protein 1; p53-inducible p53DINP1; p53DINP1; SIP TP53DINP1 TP53INP1A TP53INP1B Teap p53DINP1 p53-dependent damage-inducible nuclear protein 1 p53-inducible p53DINP1 stress-induced protein tumor protein p53-inducible nuclear protein 1; Stress-induced protein; Tumor protein p53-inducible nuclear protein 1
Gene Aliases: P53DINP1; SIP; Teap; TP53DINP1; TP53INP1; TP53INP1A; TP53INP1B
UniProt ID: (Human) Q96A56
Entrez Gene ID: (Human) 94241
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.