Novus Biologicals
Manufacturer Code:NBP176638SS
Catalog # NBP176638SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised with a synthetic peptide corresponding to 14 amino acids near the amino terminus of human p53DINP1. The immunogen is located within the first 50 amino acids of p53DINP1. Amino Acid Squence: MFQYLNKMFVGEVS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
Protein Aliases: DKFZp434M1317 FLJ22139 p53DINP1 P53DINP1p53-dependent damage-inducible nuclear protein 1 p53-inducible p53DINP1 SIPStress-induced protein Teap TP53DINP1 TP53INP1A TP53INP1B tumor protein p53 inducible nuclear protein 1 tumor protein p53-inducible nuclear protein 1; p53-dependent damage-inducible nuclear protein 1; p53-inducible p53DINP1; p53DINP1; Stress-induced protein; Tumor protein p53-inducible nuclear protein 1
Gene Aliases: P53DINP1; SIP; Teap; TP53DINP1; TP53INP1; TP53INP1A; TP53INP1B
UniProt ID: (Human) Q96A56
Entrez Gene ID: (Human) 94241
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.