Invitrogen
Manufacturer Code:MA512557
Catalog # PIMA512557
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | DO-7 |
Immunogen | Recombit human wild-type p53 protein expressed in E. coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Antigen NY-CO-13; antigen NY-CO-13 cellular tumor antigen p53 p53 tumor suppressor phosphoprotein p53 transformation-related protein 53 TP53; Cellular tumor antigen p53; mutant tumor protein 53; p53; p53 tumor suppressor; p53 tumor suppressor phosphoprotein; Phosphoprotein p53; transformation-related protein 53; tumor protein 53; tumor protein p53 (Li-Fraumeni syndrome); Tumor suppressor p53; tumor supressor p53
Gene Aliases: BCC7; LFS1; P53; TP53; TRP53
UniProt ID: (Bovine) P67939, (Human) P04637
Entrez Gene ID: (Bovine) 281542, (Human) 7157
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.