Invitrogen
Manufacturer Code:MA514067
Catalog # PIMA514067
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Type | Antibody |
Clone | DO-7 BP53-12 |
Immunogen | Recombit human p53 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Antibody Cocktail
For Research Use Only
Protein Aliases: Antigen NY-CO-13; antigen NY-CO-13 cellular tumor antigen p53 p53 tumor suppressor phosphoprotein p53 transformation-related protein 53 TP53; Cellular tumor antigen p53; mutant p53; mutant tumor protein 53; P53; p53 cellular tumor antigen; p53 tumor suppressor; Phosphoprotein p53; transformation-related protein 53; tumor protein 53; Tumor suppressor p53; tumor supressor p53
Gene Aliases: bbl; BCC7; bfy; bhy; EGK_08142; LFS1; p44; P53; TP53; TRP53
UniProt ID: (Human) P04637, (Rhesus monkey) P56424, (Mouse) P02340
Entrez Gene ID: (Human) 7157, (Rhesus monkey) 716170, (Mouse) 22059
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.