Invitrogen
Manufacturer Code:MA535658
Catalog # PIMA535658
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1206 |
Immunogen | A synthesized peptide derived from human NCF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 47 kDa autosomal chronic granulomatous disease protein; 47 kDa autosomal chronic granulomatous disease protein 47 kDa neutrophil oxidase factor Ncf1 NCF-1 NCF1A NCF-47K neutrop Neutrophil cytosol factor 1 Neutrophil NADPH oxidase factor 1 Nox organizer 2 NOXO2 Nox-organizing protein 2 P47 phox p47<phox> p47phox p47-phox SH3 and PX domain-containing protein 1A SH3PXD1A; 47 kDa neutrophil oxidase factor; NADPH oxidase organizer 2; NCF-1; NCF-47K; Neutrophil cytosol factor 1; neutrophil cytosolic factor 1, (chronic granulomatous disease, autosomal 1); Neutrophil NADPH oxidase factor 1; Nox organizer 2; Nox-organizing protein 2; p47-phox; SH3 and PX domain-containing protein 1A
Gene Aliases: NCF1; NCF1A; NOXO2; p47phox; SH3PXD1A
UniProt ID: (Human) P14598
Entrez Gene ID: (Human) 653361
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.