Invitrogen
Manufacturer Code:PA527821
Catalog # PIPA527821
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 258 of Human NCF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 47 kDa autosomal chronic granulomatous disease protein; 47 kDa neutrophil oxidase factor; NADPH oxidase organizer 2; NCF-1; NCF-47K; NCF1A NOXO2 SH3PXD1A p47phox 47 kDa autosomal chronic granulomatous disease protein 47 kDa neutrophil oxidase factor NADPH oxidase organizer 2 NCF-1 NCF-47K SH3 and PX domain-containing protein 1A neutrophil NADPH oxidase factor 1 neutrophil cytosol factor 1 neutrophil cytosolic fact; Neutrophil cytosol factor 1; neutrophil cytosolic factor 1, (chronic granulomatous disease, autosomal 1); Neutrophil NADPH oxidase factor 1; Nox organizer 2; Nox-organizing protein 2; p47-phox; SH3 and PX domain-containing protein 1A
Gene Aliases: NCF1; NCF1A; NOXO2; p47phox; SH3PXD1A
UniProt ID: (Human) P14598
Entrez Gene ID: (Human) 653361
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.