Invitrogen
Manufacturer Code:PA530780
Catalog # PIPA530780
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 136 and 430 of Human p400 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAG repeat protein 32; CAGH32 P400 TNRC12 CAG repeat protein 32 E1A-binding protein p400 domino homolog hDomino p400 SWI2/SNF2-related protein p400 kDa SWI2/SNF2-related protein trinucleotide repeat containing 12 trinucleotide repeat-containing gene 12 protein; Domino homolog; E1A-binding protein p400; hDomino; p400 kDa SWI2/SNF2-related protein; p400 SWI2/SNF2-related protein; trinucleotide repeat containing 12; Trinucleotide repeat-containing gene 12 protein
Gene Aliases: CAGH32; EP400; KIAA1498; KIAA1818; P400; TNRC12
UniProt ID: (Human) Q96L91
Entrez Gene ID: (Human) 57634
Molecular Function:
DNA binding protein
DNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.