Invitrogen
Manufacturer Code:PA5103024
Catalog # PIPA5103024
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CDK5R2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B230310J22Rik CDK5 activator 2 CDK5R2 cyclin dependent kinase 5 regulatory subunit 2 cyclin-dependent kinase 5 activator 2 cyclin-dependent kinase 5 activator isoform p39i Cyclin-dependent kinase 5 regulatory subunit 2 cyclin-dependent kinase 5 regulatory subunit 2 (p39) NCK5AI neuronal CDK5 activator isoform p39 p39I p39nck5ai RGD1562530; CDK5 activator 2; Cyclin-dependent kinase 5 activator 2; cyclin-dependent kinase 5 activator isoform p39i; Cyclin-dependent kinase 5 regulatory subunit 2; cyclin-dependent kinase 5, regulatory subunit 2 (p39); neuronal CDK5 activator isoform; p39; p39I
Gene Aliases: B230310J22Rik; CDK5R2; NCK5AI; P39; p39nck5ai; RGD1562530
UniProt ID: (Human) Q13319, (Mouse) O35926
Entrez Gene ID: (Human) 8941, (Mouse) 12570, (Rat) 501164
Molecular Function:
enzyme modulator
kinase activator
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.