Invitrogen
Manufacturer Code:PA570213
Catalog # PIPA570213
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of rat p38 MAPK beta |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MAP kinase 11; MAP kinase p38 beta; MAPK 11; mitogen activated protein kinase 11; Mitogen-activated protein kinase 11; Mitogen-activated protein kinase p38 beta; mitogen-activated protein kinase p38-2; p38-2; P38B P38BETA2 PRKM11 SAPK2 SAPK2B p38-2 p38Beta MAP kinase 11 MAP kinase p38 beta MAPK 11 mitogen-activated protein kinase p38 beta mitogen-activated protein kinase p38-2 stress-activated protein kinase 2 stress-activated protein kinase 2b stress-activated protein kinase-2 stress-activated protein kina; protein kinase, mitogen activated kinase, 11, p38beta; SAPK2b; Stress-activated protein kinase 2b; stress-activated protein kinase-2; stress-activated protein kinase-2b
Gene Aliases: MAPK11; p38-2; P38B; p38Beta; P38BETA2; PRKM11; SAPK2; SAPK2B
UniProt ID: (Human) Q15759, (Mouse) Q9WUI1
Entrez Gene ID: (Horse) 100056278, (Human) 5600, (Bovine) 618906, (Rat) 689314, (Mouse) 19094
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.