Invitrogen
Manufacturer Code:MA538272
Catalog # PIMA538272
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | HL1006 |
Immunogen | Full length human p38 MAPK recombinant protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Crk1 CSAID-binding protein Csaids binding protein CSBP Csbp1 Csbp2 CSPB1 cytokine suppressive anti-inflammatory drug binding protein cytokine suppressive anti-inflammatory drug binding protein 1 cytokine suppressive anti-inflammatory drug-binding protein cytokine-supressive anti-inflammatory drug binding protein EXIP Hog MAP kinase 14 MAP kinase 2 MAP kinase Mxi2 MAP kinase p38 alpha MAPK 14 Mapk14 MAX-interacting protein 2 mitogen activated protein kinase 14 mitogen-activated protein kinase 14 mitogen-activated protein kinase 14A Mitogen-activated protein kinase p38 alpha Mxi2 p38 p38 alpha p38 MAP kinase p38 MAP kinase alpha p38 MAPK p38 mitogen activated protein kinase p38a p38ALPHA p38-alpha p38alpha Exip p38Hog p38MAPK Prkm14 PRKM15 reactive kinase RK Sapk2A Stress-activated protein kinase 2a tRNA synthetase cofactor p38; CSAID-binding protein; Csaids binding protein; cytokine suppressive anti-inflammatory drug binding protein; Cytokine suppressive anti-inflammatory drug-binding protein; MAP kinase 14; MAP kinase MXI2; MAP kinase p38 alpha; MAX-interacting protein 2; Mitogen-activated protein kinase 14; Mitogen-activated protein kinase p38 alpha; p38 MAP kinase; p38 mitogen activated protein kinase; p38alpha Exip; SAPK2a; Stress-activated protein kinase 2a
Gene Aliases: CSBP; CSBP1; CSBP2; CSPB1; EXIP; MAPK14; MXI2; p38; p38ALPHA; PRKM14; PRKM15; RK; SAPK2A
UniProt ID: (Human) Q16539
Entrez Gene ID: (Human) 1432
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.