Novus Biologicals
Manufacturer Code:NB11096875UV
Catalog # NBT96875UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 9F12 |
Immunogen | Full length recombinant protein expressed in E.coli cells |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: CSAID-binding protein; CSAID-binding protein Csaids binding protein CSBP CSBP1MAX-interacting protein 2 CSBP2 CSPB1 cytokine suppressive inflammatory drug binding protein Cytokine suppressive inflammatory drug-binding protein EC 2.7.11 EC 2.7.11.24 EXIP MAP kinase 14 MAP kinase MXI2 MAP kinase p38 alpha MAPK 14 mitogen-activated protein kinase 14 Mitogen-activated protein kinase p38 alpha Mxi2 p38 p38 MAP kinase p38 mitogen activated protein kinase p38ALPHA p38alpha Exip PRKM14 PRKM15 RK SAPK2A stress-activated protein kinase 2A; Csaids binding protein; cytokine suppressive anti-inflammatory drug binding protein; Cytokine suppressive anti-inflammatory drug-binding protein; MAP kinase 14; MAP kinase MXI2; MAP kinase p38 alpha; MAX-interacting protein 2; Mitogen-activated protein kinase 14; Mitogen-activated protein kinase p38 alpha; p38 MAP kinase; p38 mitogen activated protein kinase; p38alpha Exip; SAPK2a; Stress-activated protein kinase 2a
Gene Aliases: CSBP; CSBP1; CSBP2; CSPB1; EXIP; MAPK14; MXI2; p38; p38ALPHA; PRKM14; PRKM15; RK; SAPK2A
UniProt ID: (Human) Q16539
Entrez Gene ID: (Human) 1432
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.