Invitrogen
Manufacturer Code:MA514834
Catalog # PIMA514834
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | B.33.3 |
Immunogen | Synthetic peptide from the carboxy terminus of human p35 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CDK5 activator 1; CDK5 activator 1 TPKII regulatory subunit cyclin-dependent kinase 5 activator 1 cyclin-dependent kinase 5 regulatory subunit 1 neuronal CDK5 activator regulatory partner for CDK5 kinase tau protein kinase II 23kDa subunit; Cyclin-dependent kinase 5 activator 1; Cyclin-dependent kinase 5 activator 1, p25; Cyclin-dependent kinase 5 activator 1, p35; Cyclin-dependent kinase 5 regulatory subunit 1; cyclin-dependent kinase 5, regulatory subunit (p35) 1; cyclin-dependent kinase 5, regulatory subunit 1 (p35); neuronal CDK5 activator; p23; p25; p35; regulatory partner for CDK5 kinase; Tau protein kinase II 23 kDa subunit; tau protein kinase II 23kDa subunit; TPKII regulatory subunit
Gene Aliases: CDK5P35; CDK5R; CDK5R1; D11Bwg0379e; NCK5A; p23; p25; p35; p35nck5a
UniProt ID: (Human) Q15078, (Rat) P61810, (Mouse) P61809
Entrez Gene ID: (Human) 8851, (Rat) 116671, (Mouse) 12569
Molecular Function:
enzyme modulator
kinase activator
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.