Novus Biologicals
Manufacturer Code:NBP2444970.2MG
Catalog # NBP24449702
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | DCS-72.F6 + KIP1/769 |
Immunogen | Mouse recombinant p27 protein (DCS-72.F6) Recombinant human CDKN1B protein (KIP1/769) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CDKN4 cyclin-dependent kinase inhibitor 1B cyclin-dependent kinase inhibitor 1B (p27 Kip1) Cyclin-dependent kinase inhibitor p27 KIP1P27KIP1 MEN1B MEN4 p27Kip1; Cyclin-dependent kinase inhibitor 1B; cyclin-dependent kinase inhibitor 1B (p27, Kip1); Cyclin-dependent kinase inhibitor p27; p27Kip1
Gene Aliases: CDKN1B; CDKN4; KIP1; MEN1B; MEN4; P27KIP1
UniProt ID: (Human) P46527
Entrez Gene ID: (Human) 1027
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.