Invitrogen
Manufacturer Code:PA517946
Catalog # PIPA517946
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (EQTPKKPGLRRRQT) corresponding to the C-terminus amino acids of CDKN1B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CDKN1B P27KIP1 cyclin-dependent kinase inhibitor 1B (p27 Kip1) KIP1 CDKN4 cyclin-dependent kinase inhibitor 1B p27(KIP1) KIP1 p27 MEN1B MEN4 P27KIP1; Cyclin-dependent kinase inhibitor 1B; cyclin-dependent kinase inhibitor 1B (p27, Kip1); Cyclin-dependent kinase inhibitor p27; p27Kip1
Gene Aliases: CDKN1B; CDKN4; KIP1; MEN1B; MEN4; P27KIP1
UniProt ID: (Human) P46527
Entrez Gene ID: (Human) 1027
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.