R&D Systems
Manufacturer Code:IC1047N100UG
Catalog # IC1047N100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 195720 |
Immunogen | <EM>E. coli</EM>-derived recombinant human p21/CIP1/CDKN1A Ser2-Pro164 Accession # P38936 |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: CAP20cyclin-dependent kinase inhibitor 1 CDK-interacting protein 1 CDKN1A CDKN1melanoma differentiation associated protein 6 CIP1 CIP1WAF1CDK-interaction protein 1 cyclin-dependent kinase inhibitor 1A (p21 Cip1) MDA6 MDA-6 Melanoma differentiation-associated protein 6 p21 p21CIP1 p21Cip1/Waf1 PIC1 SDI1DNA synthesis inhibitor wild-type p53-activated fragment 1; CDK-interacting protein 1; CDK-interaction protein 1; Cyclin-dependent kinase inhibitor 1; cyclin-dependent kinase inhibitor 1A (p21, Cip1); DNA synthesis inhibitor; MDA-6; melanoma differentiation associated protein 6; Melanoma differentiation-associated protein 6; p21; wild-type p53-activated fragment 1
Gene Aliases: CAP20; CDKN1; CDKN1A; CIP1; MDA-6; MDA6; P21; p21CIP1; PIC1; SDI1; WAF1
UniProt ID: (Human) P38936
Entrez Gene ID: (Human) 1026
Molecular Function: enzyme modulator kinase inhibitor kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.