Novus Biologicals
Manufacturer Code:NBP286741
Catalog # NB034363
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of rat p19ARF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alternative reading frame; alternative reading frame ARF ARF-INK4a CDK4I CDKN2A Cyclin-dependent kinase 4 inhibitor A cyclin-dependent kinase inhibitor 2A cyclin-dependent kinase inhibitor 2A (p16 inhibits CDK4) cyclin-dependent kinase inhibitor 2A isoform 3 cyclin-dependent kinase inhibitor 2A isoforms 1/2 cyclin-dependent kinase inhibitor protein INK4 INK4a Ink4a/Arf INK4a-ARF mitochondrial smARF&prime MTS1 p14 p14ARF P16 p16(INK4a) p16-INK4 p16INK4a p16-INK4a P19 p19<ARF> p19ARF Pctr1; ARF; CDK4I; cdkn2a; Cyclin-dependent kinase 4 inhibitor A; Cyclin-dependent kinase inhibitor 2A; cyclin-dependent kinase inhibitor 2A (p16, inhibits CDK4); cyclin-dependent kinase inhibitor 2A, isoform 3; cyclin-dependent kinase inhibitor 2A, isoforms 1/2; cyclin-dependent kinase inhibitor protein; mitochondrial smARF; p16-INK4; p16-INK4a; p19ARF; Tumor suppressor ARF
Gene Aliases: Arf; ARF-INK4a; Cdkn2a; INK4a-ARF; Ink4a/Arf; MTS1; p16; p16(INK4a); P16ink4a; p19<ARF>; p19ARF; Pctr1
UniProt ID: (Mouse) Q78E39
Entrez Gene ID: (Mouse) 12578
Molecular Function:
enzyme modulator
kinase inhibitor
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.