Novus Biologicals
Manufacturer Code:NB200174AF405
Catalog # NB200174W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 5-C3-1 |
Immunogen | A synthetic peptide containing amino acids 54-75 of the murine p19ARF. [UniProt# Q64364] |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Alternative reading frame; ARF; ARF CDK4 inhibitor p16-INK4 CDK4IP14ARF CDKN2 cell cycle negative regulator beta CMM2P16-INK4A Cyclin-dependent kinase 4 inhibitor A cyclin-dependent kinase inhibitor 2A cyclin-dependent kinase inhibitor 2A (melanoma p16 inhibits CDK4) INK4 INK4A MLMP16INK4 MTS-1 MTS1P14 Multiple tumor suppressor 1 p14 p14ARF p16 p16-INK4 p16INK4a p16-INK4a P19 p19ARF TP16; CDK4I; cdkn2a; Cyclin-dependent kinase 4 inhibitor A; Cyclin-dependent kinase inhibitor 2A; cyclin-dependent kinase inhibitor 2A (p16, inhibits CDK4); cyclin-dependent kinase inhibitor 2A, isoform 3; cyclin-dependent kinase inhibitor 2A, isoforms 1/2; cyclin-dependent kinase inhibitor protein; mitochondrial smARF; p16-INK4; p16-INK4a; p19ARF; Tumor suppressor ARF
Gene Aliases: Arf; ARF-INK4a; Cdkn2a; INK4a-ARF; Ink4a/Arf; MTS1; p16; p16(INK4a); P16ink4a; p19<ARF>; p19ARF; Pctr1
UniProt ID: (Mouse) Q78E39
Entrez Gene ID: (Mouse) 12578
Molecular Function:
enzyme modulator
kinase inhibitor
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.