Novus Biologicals
Manufacturer Code:NBP2842060.1ML
Catalog # NB032134
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human CDKN2A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alternative reading frame; ARF; ARF CDK4 inhibitor p16-INK4 CDK4IP14ARF CDKN2 cell cycle negative regulator beta CMM2P16-INK4A Cyclin-dependent kinase 4 inhibitor A cyclin-dependent kinase inhibitor 2A cyclin-dependent kinase inhibitor 2A (melanoma p16 inhibits CDK4) INK4 INK4A MLMP16INK4 MTS-1 MTS1P14 Multiple tumor suppressor 1 p14 p14ARF p16 p16-INK4 p16INK4a p16-INK4a P19 p19ARF TP16; CDK4 inhibitor p16-INK4; CDK4I; cell cycle negative regulator beta; Cyclin-dependent kinase 4 inhibitor A; Cyclin-dependent kinase inhibitor 2A; cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4); MTS-1; Multiple tumor suppressor 1; p14ARF; p16-INK4; p16-INK4a; Tumor suppressor ARF
Gene Aliases: ARF; CDK4I; CDKN2; CDKN2A; CMM2; INK4; INK4A; MLM; MTS-1; MTS1; P14; P14ARF; P16; P16-INK4A; P16INK4; P16INK4A; P19; P19ARF; TP16
UniProt ID: (Human) P42771
Entrez Gene ID: (Human) 1029
Molecular Function: enzyme modulator kinase inhibitor kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.