Invitrogen
Manufacturer Code:MA537841
Catalog # PIMA537841
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0661 |
Immunogen | A synthesized peptide derived from human CDKN2A/p16INK4a |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Alternative reading frame; Alternative reading frame Arf ARF-INK4a CDK4 inhibitor p16-INK4 CDK4I CDKN2 CDKN2A cdkn2a {ECO:0000312 cell cycle inhibitor cell cycle negative regulator beta cell cycle regulator CMM2 cyclin dependent kinase inhibitor 2A Cyclin dependent kinase inhibitor 2A (p16 inhibits CDK4) Cyclin-dependent kinase 4 inhibitor A cyclin-dependent kinase inhibitor 2A cyclin-dependent kinase inhibitor 2A (melanoma p16 inhibits CDK4) cyclin-dependent kinase inhibitor 2A (p16 inhibits CDK4) cyclin-dependent kinase inhibitor 2a p16Ink4a cyclin-dependent kinase inhibitor 2a p19Arf cyclin-dependent kinase inhibitor 2A isoform 1 cyclin-dependent kinase inhibitor 2A isoform 2 cyclin-dependent kinase inhibitor 2A isoform 3 cyclin-dependent kinase inhibitor 2A isoforms 1/2 cyclin-dependent kinase inhibitor protein EMBL:AAL76336.1 ECO:0000312 EMBL:AAL76338.1 ECO:0000312 HGNC:1787 inhibits CDK4 INK4 INK4A Ink4a/Arf INK4a-ARF isoforms 1/2/3 mitochondrial smARF MLM MTS1 MTS-1 Multiple tumor suppressor 1 P14 P14 P16 P14ARF p16 p16(INK4a) p16Cdkn2a P16INK4 p16-INK4 P16INK4A p16-INK4a P19 P19 TP16 p19<ARF> p19ARF Pctr1 RGD:2323} TP16 Tumor suppressor ARF; ARF; CDK4 inhibitor p16-INK4; CDK4I; cell cycle negative regulator beta; Cyclin-dependent kinase 4 inhibitor A; Cyclin-dependent kinase inhibitor 2A; cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4); MTS-1; Multiple tumor suppressor 1; p14ARF; p16-INK4; p16-INK4a; Tumor suppressor ARF
Gene Aliases: ARF; CDK4I; CDKN2; CDKN2A; CMM2; INK4; INK4A; MLM; MTS-1; MTS1; P14; P14ARF; P16; P16-INK4A; P16INK4; P16INK4A; P19; P19ARF; TP16
UniProt ID: (Human) P42771
Entrez Gene ID: (Human) 1029
Molecular Function:
enzyme modulator
kinase inhibitor
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.