Proteintech
Manufacturer Code:551821AP150UL
Catalog # 50-173-6040
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 150 kDa dynein-associated polypeptide; DAP 150 DCTN1 DP 150 Dynactin subunit 1 HMN7B P135 p150 glued; DAP-150; DP-150; dynactin 1 (p150, glued homolog, Drosophila); dynactin 1, retrograde axonal transport; Dynactin subunit 1; p135; p150-glued
Gene Aliases: AL022633; DAP-150; DCTN1; DP-150; Glued; P135; p150; p150<glued>
UniProt ID: (Human) Q14203, (Rat) P28023, (Mouse) O08788
Entrez Gene ID: (Human) 1639, (Dog) 483106, (Rat) 29167, (Mouse) 13191
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.