Invitrogen
Manufacturer Code:MA535623
Catalog # PIMA535623
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1144 |
Immunogen | A synthesized peptide derived from human CDKN2A/p14ARF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Alternative reading frame; Alternative reading frame ARF ARF-INK4a CDK4I CDKN2 Cdkn2a CMM2 Cyclin-dependent kinase 4 inhibitor A Cyclin-dependent kinase inhibitor 2A inhibits CDK4 INK4 INK4A Ink4a/Arf INK4a-ARF MLM MTS1 MTS-1 Multiple tumor suppressor 1 P14 p14 p16 p14ARF p16 p16(INK4a) p16Cdkn2a P16INK4 p16-INK4 P16ink4a p16-INK4a P19 P19 TP16 p19<ARF> p19ARF Pctr1 TP16 Tumor suppressor ARF; ARF; CDK4 inhibitor p16-INK4; CDK4I; cell cycle negative regulator beta; Cyclin-dependent kinase 4 inhibitor A; Cyclin-dependent kinase inhibitor 2A; cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4); MTS-1; Multiple tumor suppressor 1; p14ARF; p16-INK4; p16-INK4a; Tumor suppressor ARF
Gene Aliases: ARF; CDK4I; CDKN2; CDKN2A; CMM2; INK4; INK4A; MLM; MTS-1; MTS1; P14; P14ARF; P16; P16-INK4A; P16INK4; P16INK4A; P19; P19ARF; TP16
UniProt ID: (Human) P42771
Entrez Gene ID: (Human) 1029
Molecular Function:
enzyme modulator
kinase inhibitor
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.