Invitrogen
Manufacturer Code:PA585607
Catalog # PIPA585607
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human CDKN2A/p14ARF. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alternative reading frame; ARF; CDK4 inhibitor p16-INK4; CDK4I; cell cycle negative regulator beta; Cyclin-dependent kinase 4 inhibitor A; Cyclin-dependent kinase inhibitor 2A; cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4); cyclin-dependent kinase inhibitor 2A Antibody P16INK4 Antibody TP16 Antibody CDKN2 Antibody MTS1 Antibody P16 Antibody P14ARF Antibody CDKN2A Antibody CMM2 Antibody P16INK4A Antibody MLM Antibody ARF Antibody INK4 Antibody P19ARF Antibody P19 Antibody INK4A Antibody P16-INK4A Antibody CDK4I Antibody P14 Antibody MTS-1 Antibody; MTS-1; Multiple tumor suppressor 1; p14ARF; p16-INK4; p16-INK4a; Tumor suppressor ARF
Gene Aliases: ARF; CDK4I; CDKN2; CDKN2A; CMM2; INK4; INK4A; MLM; MTS-1; MTS1; P14; P14ARF; P16; P16-INK4A; P16INK4; P16INK4A; P19; P19ARF; TP16
UniProt ID: (Human) P42771
Entrez Gene ID: (Human) 1029
Molecular Function: enzyme modulator kinase inhibitor kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.