Invitrogen
Manufacturer Code:PA578381
Catalog # PIPA578381
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of mouse p130Cas. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI385681 Bcar1 Cas Crkas breast cancer anti-estrogen resistance 1; Breast cancer anti-estrogen resistance protein 1; CRK-associated substrate; p130 Cas; p130cas; v-crk-associated tyrosine kinase substrate
Gene Aliases: AI385681; Bcar1; Cas; Crkas; P130CAS
UniProt ID: (Mouse) Q61140, (Rat) Q63767
Entrez Gene ID: (Mouse) 12927, (Rat) 25414
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.