Novus Biologicals
Manufacturer Code:NBP252551
Catalog # NBP252551
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2C8A8 |
Immunogen | Purified recombinant fragment of human RBL2 (AA: 939-1139) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 130 kDa retinoblastoma-associated protein; FLJ26459 P130 PRB2 RB2 RBR-2 retinoblastoma-like 2 (p130) retinoblastoma-like protein 2130 kDa retinoblastoma-associated protein Retinoblastoma-related protein 2; p130; PRB2; RBR-2; retinoblastoma-like 2; Retinoblastoma-like protein 2; Retinoblastoma-related protein 2
Gene Aliases: P130; RB2; RBL2
UniProt ID: (Human) Q08999
Entrez Gene ID: (Human) 5934
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.