Novus Biologicals
Manufacturer Code:NB6001541
Catalog # NB6001541
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 130P215 |
Immunogen | Synthetic peptide: QCPELMMDRHLDQLLMCAIYVMAKVTKEDKSFQNIM corresponding to amino acids 878-913 of Human p130. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 130 kDa retinoblastoma-associated protein; FLJ26459 P130 PRB2 RB2 RBR-2 retinoblastoma-like 2 (p130) retinoblastoma-like protein 2130 kDa retinoblastoma-associated protein Retinoblastoma-related protein 2; p130; PRB2; RBR-2; retinoblastoma-like 2; Retinoblastoma-like protein 2; Retinoblastoma-related protein 2
Gene Aliases: P130; RB2; RBL2
UniProt ID: (Human) Q08999
Entrez Gene ID: (Human) 5934
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.