Invitrogen
Manufacturer Code:PA578147
Catalog # PIPA578147
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human p130. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 130 kDa retinoblastoma-associated protein; P130; PPAR-alpha-interacting complex protein 128; PRB2; PRIC128; RBR-2; retinoblastoma-like 2; retinoblastoma-like 2 (p130) Rb2 P130 RBL2; Retinoblastoma-like protein 2; Retinoblastoma-related protein 2
Gene Aliases: P130; PRB2; RB2; RBL2; RBR-2
UniProt ID: (Human) Q08999, (Mouse) Q64700, (Rat) O55081
Entrez Gene ID: (Human) 5934, (Mouse) 19651, (Rat) 81758
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.